siRNA Details
| Gene Symbol: Ggcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031756.1 | NP_113944.1 | 4 | 486 | ||
| GenBank mRNA | AF065387.1 | - | 4 | 486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggcx| Gene Symbol: Ggcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031756.1 | NP_113944.1 | 4 | 486 | ||
| GenBank mRNA | AF065387.1 | - | 4 | 486 | ||