siRNA Details
| Gene Symbol: Ctbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053335.1 | NP_445787.1 | 1 | 1718 | ||
| GenBank mRNA | AF222712.1 | - | 1 | 1718 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctbp2| Gene Symbol: Ctbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053335.1 | NP_445787.1 | 1 | 1718 | ||
| GenBank mRNA | AF222712.1 | - | 1 | 1718 | ||