siRNA Details
| Gene Symbol: Chrna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057184.1 | NP_476532.1 | 5 | 1133 | ||
| GenBank mRNA | L08227.1 | - | 5 | 1133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna6| Gene Symbol: Chrna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057184.1 | NP_476532.1 | 5 | 1133 | ||
| GenBank mRNA | L08227.1 | - | 5 | 1133 | ||