siRNA Details
| Gene Symbol: Ager | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053336.2 | NP_445788.2 | 8, 9 | 1000 | ||
| GenBank mRNA | BC084697.1 | - | 8, 9 | 982 | 7121345 | |
| GU164716.1 | - | 7, 8 | 1167 | |||
| GU164718.1 | - | 4, 5 | 793 | |||
| GU164720.1 | - | 4, 5 | 344 | |||
| L33413.1 | - | 8, 9 | 1000 | |||

Ager
Genomic Map