siRNA Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031062.2 | NP_112324.1 | 7, 8 | 942 | ||
| GenBank mRNA | BC081784.1 | - | 7, 8 | 927 | 7135885 | |
| U53706.1 | - | 7, 8 | 943 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mvd| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031062.2 | NP_112324.1 | 7, 8 | 942 | ||
| GenBank mRNA | BC081784.1 | - | 7, 8 | 927 | 7135885 | |
| U53706.1 | - | 7, 8 | 943 | |||