siRNA Details
| Gene Symbol: Fez1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031066.1 | NP_112328.1 | 5, 6 | 823 | ||
| GenBank mRNA | BC087740.1 | - | 5, 6 | 823 | 7314065 | |
| FQ212529.1 | - | 3, 4 | 569 | |||
| U48249.2 | - | 5, 6 | 786 | |||
| U63740.1 | - | 5, 6 | 823 | |||

Fez1
Genomic Map