siRNA Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276711.1 | NP_001263640.1 | 8 | 868 | ||
| XM_006233358.2 | XP_006233420.1 | 6 | 524 | |||
| XM_006233360.2 | XP_006233422.1 | 6 | 524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfkb1| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276711.1 | NP_001263640.1 | 8 | 868 | ||
| XM_006233358.2 | XP_006233420.1 | 6 | 524 | |||
| XM_006233360.2 | XP_006233422.1 | 6 | 524 | |||