siRNA Details
Gene Symbol: Nfkb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276711.1 | NP_001263640.1 | 24, 25 | 3115 | ||
XM_006233358.2 | XP_006233420.1 | 22, 23 | 2771 | |||
GenBank mRNA | FQ232433.1 | - | 7, 8 | 853 | ||
L26267.1 | - | 10, 11 | 1411 |
