siRNA Details
| Gene Symbol: Ntf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270868.1 | NP_001257797.1 | 3 | 310 | ||
| NM_001270869.1 | NP_001257798.1 | 2 | 272 | |||
| NM_001270870.1 | NP_001257799.1 | 2 | 244 | |||
| NM_031073.3 | NP_112335.3 | 3 | 354 | |||
| XM_008763294.1 | XP_008761516.1 | 2 | 326 | |||
| XM_008763295.1 | XP_008761517.1 | 2 | 306 | |||
| GenBank mRNA | BC070504.1 | - | 3 | 354 | 7097740 | |
| EV778881.1 | - | 2 | 261 | |||
| FM074856.1 | - | 3 | 309 | |||
| FM101680.1 | - | 2 | 271 | |||
| M34643.1 | - | 2 | 105 | |||

Ntf3
Genomic Map