siRNA Details
| Gene Symbol: Pcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031076.1 | NP_112338.1 | 12, 13 | 2268 | ||
| XM_006253183.2 | XP_006253245.1 | 14, 15 | 2765 | |||
| XM_006253184.2 | XP_006253246.1 | 15, 16 | 2869 | |||
| XM_006253185.2 | XP_006253247.1 | 14, 15 | 2765 | |||
| XM_006253186.2 | XP_006253248.1 | 14, 15 | 2764 | |||
| XM_006253187.2 | XP_006253249.1 | 13, 14 | 2590 | |||
| XM_006253188.2 | XP_006253250.1 | 13, 14 | 2575 | |||
| XM_008771284.1 | XP_008769506.1 | 15, 16 | 2884 | |||
| XM_008771285.1 | XP_008769507.1 | 14, 15 | 2613 | |||
| XM_008771286.1 | XP_008769508.1 | 14, 15 | 2543 | |||

Pcm1
Genomic Map