siRNA Details
| Gene Symbol: Pcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031076.1 | NP_112338.1 | 24, 25 | 4484 | ||
| XM_006253183.2 | XP_006253245.1 | 26, 27 | 4981 | |||
| XM_006253184.2 | XP_006253246.1 | 27, 28 | 5085 | |||
| XM_006253185.2 | XP_006253247.1 | 26, 27 | 4981 | |||
| XM_006253186.2 | XP_006253248.1 | 25, 26 | 4815 | |||
| XM_006253187.2 | XP_006253249.1 | 25, 26 | 4806 | |||
| XM_006253188.2 | XP_006253250.1 | 25, 26 | 4791 | |||
| XM_008771284.1 | XP_008769506.1 | 27, 28 | 5100 | |||
| XM_008771285.1 | XP_008769507.1 | 26, 27 | 4829 | |||
| XM_008771286.1 | XP_008769508.1 | 26, 27 | 4759 | |||

Pcm1
Genomic Map