siRNA Details
| Gene Symbol: Pcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031076.1 | NP_112338.1 | 20, 21 | 3631 | ||
| XM_006253183.2 | XP_006253245.1 | 22, 23 | 4128 | |||
| XM_006253184.2 | XP_006253246.1 | 23, 24 | 4232 | |||
| XM_006253185.2 | XP_006253247.1 | 22, 23 | 4128 | |||
| XM_006253186.2 | XP_006253248.1 | 22, 23 | 4127 | |||
| XM_006253187.2 | XP_006253249.1 | 21, 22 | 3953 | |||
| XM_006253188.2 | XP_006253250.1 | 21, 22 | 3938 | |||
| XM_008771284.1 | XP_008769506.1 | 23, 24 | 4247 | |||
| XM_008771285.1 | XP_008769507.1 | 22, 23 | 3976 | |||
| XM_008771286.1 | XP_008769508.1 | 22, 23 | 3906 | |||

Pcm1
Genomic Map