siRNA Details
| Gene Symbol: Cdk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004132.1 | NP_001004132.1 | 8, 9 | 805 | ||
| NM_031077.1 | NP_112339.1 | 8, 9 | 1164 | |||
| XM_006256617.2 | XP_006256679.1 | 8, 9 | 4932 | |||
| XM_006256618.2 | XP_006256680.1 | 8, 9 | 1165 | |||
| XM_006256619.2 | XP_006256681.1 | 8, 9 | 1217 | |||
| XM_008773037.1 | XP_008771259.1 | 8, 9 | 4935 | |||
| GenBank mRNA | BC078711.1 | - | 8, 9 | 805 | 7112874 | |
| U36444.1 | - | 8, 9 | 1164 | |||

Cdk16
Genomic Map