siRNA Details
| Gene Symbol: Pde2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143847.1 | NP_001137319.1 | 22 | 2039 | ||
| NM_001270604.1 | NP_001257533.1 | 22 | 1989 | |||
| NM_031079.1 | NP_112341.1 | 21 | 1864 | |||
| GenBank mRNA | U21101.1 | - | 21 | 1864 | ||

Pde2a
Genomic Map