siRNA Details
| Gene Symbol: Pde2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143847.1 | NP_001137319.1 | 27 | 2459 | ||
| NM_001270604.1 | NP_001257533.1 | 27 | 2409 | |||
| NM_031079.1 | NP_112341.1 | 26 | 2284 | |||
| GenBank mRNA | M94540.1 | - | 5 | 321 | ||
| U21101.1 | - | 26 | 2284 | |||

Pde2a
Genomic Map