siRNA Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031081.1 | NP_112343.1 | 14 | 1693 | ||
| XM_006246036.2 | XP_006246098.1 | 14 | 3545 | |||
| GenBank mRNA | Y15748.1 | - | 14 | 1693 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdpk1| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031081.1 | NP_112343.1 | 14 | 1693 | ||
| XM_006246036.2 | XP_006246098.1 | 14 | 3545 | |||
| GenBank mRNA | Y15748.1 | - | 14 | 1693 | ||