siRNA Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031081.1 | NP_112343.1 | 10 | 1075 | ||
| XM_006246036.2 | XP_006246098.1 | 10 | 2927 | |||
| XM_006246038.2 | XP_006246100.1 | 10 | 1137 | |||
| GenBank mRNA | Y15748.1 | - | 10 | 1075 | ||

Pdpk1
Genomic Map