siRNA Details
| Gene Symbol: Pi4kb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031083.1 | NP_112345.1 | 9 | 2351 | ||
| XM_008761332.1 | XP_008759554.1 | 11 | 2693 | |||
| XM_008761333.1 | XP_008759555.1 | 10 | 2501 | |||
| XM_008761334.1 | XP_008759556.1 | 10 | 2574 | |||
| XM_008761335.1 | XP_008759557.1 | 10 | 2648 | |||
| XM_008761336.1 | XP_008759558.1 | 8 | 2337 | |||
| GenBank mRNA | AY724527.1 | - | 8 | 2073 | ||
| D84667.1 | - | 9 | 2351 | |||
| FQ213038.1 | - | 1 | 14 | |||

Pi4kb
Genomic Map