siRNA Details
Gene Symbol: Pi4kb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031083.1 | NP_112345.1 | 8 | 2043 | ||
XM_008761332.1 | XP_008759554.1 | 10 | 2385 | |||
XM_008761333.1 | XP_008759555.1 | 9 | 2193 | |||
XM_008761334.1 | XP_008759556.1 | 9 | 2266 | |||
XM_008761335.1 | XP_008759557.1 | 9 | 2340 | |||
XM_008761336.1 | XP_008759558.1 | 7 | 2029 | |||
GenBank mRNA | AY724527.1 | - | 7 | 1765 | ||
D84667.1 | - | 8 | 2043 |
