siRNA Details
| Gene Symbol: Rbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031094.1 | NP_112356.1 | 3 | 456 | ||
| XM_006255199.2 | XP_006255261.1 | 3 | 510 | |||
| XM_006255200.2 | XP_006255262.1 | 3 | 511 | |||
| GenBank mRNA | D55627.1 | - | 3 | 456 | ||

Rbl2
Genomic Map