siRNA Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031097.1 | NP_112359.1 | 7, 8 | 1320 | ||
| GenBank mRNA | AY724503.1 | - | 7, 8 | 1495 | ||
| BC061718.1 | - | 7, 8 | 1353 | 5598331 | ||
| D87515.1 | - | 7, 8 | 1317 | |||
| U61696.1 | - | 7, 8 | 1320 | |||

Rnpep
Genomic Map