siRNA Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031097.1 | NP_112359.1 | 8, 9 | 1421 | ||
| GenBank mRNA | AY724503.1 | - | 8, 9 | 1596 | ||
| BC061718.1 | - | 8, 9 | 1454 | 5598331 | ||
| D87515.1 | - | 8, 9 | 1418 | |||
| U61696.1 | - | 8, 9 | 1421 | |||

Rnpep
Genomic Map