siRNA Details
Gene Symbol: Soat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031118.1 | NP_112380.1 | 16 | 1711 | ||
GenBank mRNA | D86373.1 | - | 16 | 1711 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Soat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031118.1 | NP_112380.1 | 16 | 1711 | ||
GenBank mRNA | D86373.1 | - | 16 | 1711 |