siRNA Details
| Gene Symbol: Ssr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031120.2 | NP_112382.2 | 1 | 426 | ||
| GenBank mRNA | AY724496.1 | - | 1 | 434 | ||
| BC062015.1 | - | 1 | 426 | 5621495 | ||
| Z14030.1 | - | 434 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssr3| Gene Symbol: Ssr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031120.2 | NP_112382.2 | 1 | 426 | ||
| GenBank mRNA | AY724496.1 | - | 1 | 434 | ||
| BC062015.1 | - | 1 | 426 | 5621495 | ||
| Z14030.1 | - | 434 | ||||