siRNA Details
| Gene Symbol: Ssrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031121.1 | NP_112383.1 | 6 | 1088 | ||
| XM_006234477.1 | XP_006234539.1 | 7 | 1092 | |||
| XM_008761993.1 | XP_008760215.1 | 7 | 1074 | |||
| XM_008761994.1 | XP_008760216.1 | 7 | 998 | |||
| GenBank mRNA | BC083588.1 | - | 6 | 1088 | 7132288 | |
| L08814.1 | - | 3 | 351 | |||

Ssrp1
Genomic Map