siRNA Details
| Gene Symbol: Stxbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031126.1 | NP_112388.1 | 5 | 303 | ||
| XM_006248783.2 | XP_006248845.1 | 5 | 350 | |||
| GenBank mRNA | BC097310.1 | - | 5 | 326 | ||
| U20283.1 | - | 5 | 303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stxbp2| Gene Symbol: Stxbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031126.1 | NP_112388.1 | 5 | 303 | ||
| XM_006248783.2 | XP_006248845.1 | 5 | 350 | |||
| GenBank mRNA | BC097310.1 | - | 5 | 326 | ||
| U20283.1 | - | 5 | 303 | |||