siRNA Details
| Gene Symbol: Suox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031127.3 | NP_112389.3 | 3 | 269 | ||
| XM_006240802.2 | XP_006240864.1 | 4 | 349 | |||
| XM_006240803.2 | XP_006240865.1 | 5 | 450 | |||
| GenBank mRNA | BC061991.1 | - | 3 | 269 | 5599452 | |
| L05084.1 | - | 2 | 125 | |||

Suox
Genomic Map