siRNA Details
| Gene Symbol: Serpina7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257307.2 | XP_006257369.1 | 3 | 169 | ||
| XM_006257308.2 | XP_006257370.1 | 2 | 1426 | |||
| XM_008773465.1 | XP_008771687.1 | 2 | 1424 | |||
| XM_008773466.1 | XP_008771688.1 | 3 | 162 | |||
| XM_343823.6 | XP_343824.4 | 2 | 98 | |||

Serpina7
Genomic Map