siRNA Details
| Gene Symbol: Thra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017960.1 | NP_001017960.1 | 9 | 1454 | ||
| NM_031134.2 | NP_112396.2 | 8 | 1322 | |||
| GenBank mRNA | M18028.1 | - | 9 | 1344 | ||
| M31174.1 | - | 1536 | ||||
| M31177.1 | - | 4 | 663 | |||
| X07409.1 | - | 7 | 983 | |||
| X12744.1 | - | 8 | 986 | |||

Thra
Genomic Map