siRNA Details
| Gene Symbol: Thra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017960.1 | NP_001017960.1 | 5, 6 | 805 | ||
| NM_031134.2 | NP_112396.2 | 4, 5 | 673 | |||
| GenBank mRNA | CB728734.1 | - | 1, 2 | 154 | ||
| M18028.1 | - | 5, 6 | 695 | |||
| M31174.1 | - | 887 | ||||
| M31177.1 | - | 14 | ||||
| X07409.1 | - | 3, 4 | 334 | |||
| X12744.1 | - | 4, 5 | 337 | |||

Thra
Genomic Map