siRNA Details
| Gene Symbol: Tpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031137.1 | NP_112399.1 | 1409 | |||
| GenBank mRNA | U50194.1 | - | 1409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpp2| Gene Symbol: Tpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031137.1 | NP_112399.1 | 1409 | |||
| GenBank mRNA | U50194.1 | - | 1409 | |||