siRNA Details
| Gene Symbol: Arpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031146.2 | NP_112408.1 | 4, 5 | 547 | ||
| GenBank mRNA | AF315378.1 | - | 3, 4 | 432 | ||
| BC059131.1 | - | 4, 5 | 527 | 6921520 | ||
| CO396710.1 | - | 15 | ||||
| DY310102.1 | - | 4, 5 | 547 | |||
| FQ214345.1 | - | 4, 5 | 521 | |||

Arpc1a
Genomic Map