siRNA Details
| Gene Symbol: Psmc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031149.1 | NP_112411.1 | 4 | 246 | ||
| GenBank mRNA | AB000491.1 | - | 4 | 246 | ||
| BC058462.1 | - | 4 | 242 | 6891005 | ||
| D83521.1 | - | 4 | 264 | |||
| FQ213263.1 | - | 4 | 263 | |||
| FQ213735.1 | - | 4 | 238 | |||
| FQ233831.1 | - | 4 | 249 | |||

Psmc5
Genomic Map