siRNA Details
| Gene Symbol: Cyp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031241.1 | NP_112520.1 | 1 | 254 | ||
| GenBank mRNA | AB009686.1 | - | 1 | 254 | ||
| AB018596.1 | - | 1 | 221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp8b1| Gene Symbol: Cyp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031241.1 | NP_112520.1 | 1 | 254 | ||
| GenBank mRNA | AB009686.1 | - | 1 | 254 | ||
| AB018596.1 | - | 1 | 221 | |||