siRNA Details
| Gene Symbol: Pias2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053337.1 | NP_445789.1 | 953 | |||
| GenBank mRNA | AF044058.1 | - | 953 | |||
| BC078775.1 | - | 952 | 7111628 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pias2| Gene Symbol: Pias2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053337.1 | NP_445789.1 | 953 | |||
| GenBank mRNA | AF044058.1 | - | 953 | |||
| BC078775.1 | - | 952 | 7111628 | |||