siRNA Details
Gene Symbol: Pias2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053337.1 | NP_445789.1 | 1633 | |||
GenBank mRNA | AF044058.1 | - | 1633 | |||
BC078775.1 | - | 1632 | 7111628 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pias2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053337.1 | NP_445789.1 | 1633 | |||
GenBank mRNA | AF044058.1 | - | 1633 | |||
BC078775.1 | - | 1632 | 7111628 |