siRNA Details
| Gene Symbol: Tfam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031326.1 | NP_112616.1 | 7 | 826 | ||
| GenBank mRNA | AB014089.1 | - | 7 | 826 | ||
| AF246196.1 | - | 6 | 603 | |||
| AF377866.1 | - | 7 | 833 | |||
| AJ312746.1 | - | 7 | 826 | |||
| BC062022.1 | - | 7 | 828 | 5623491 | ||
| FQ219683.1 | - | 4 | 384 | |||

Tfam
Genomic Map