siRNA Details
| Gene Symbol: Timeless | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031340.1 | NP_112630.1 | 22 | 2758 | ||
| GenBank mRNA | AB019576.1 | - | 22 | 2758 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timeless| Gene Symbol: Timeless | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031340.1 | NP_112630.1 | 22 | 2758 | ||
| GenBank mRNA | AB019576.1 | - | 22 | 2758 | ||