siRNA Details
| Gene Symbol: Slc7a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031341.1 | NP_112631.1 | 8 | 1391 | ||
| XM_006252020.2 | XP_006252082.1 | 8 | 1327 | |||
| XM_006252021.2 | XP_006252083.1 | 11 | 1796 | |||
| XM_006252023.2 | XP_006252085.1 | 8 | 1332 | |||
| XM_006252024.2 | XP_006252086.1 | 8 | 2021 | |||
| XM_008770705.1 | XP_008768927.1 | 10 | 1739 | |||
| GenBank mRNA | AB020520.1 | - | 8 | 1391 | ||
| AF200684.1 | - | 7 | 1199 | |||
| BC091142.1 | - | 8 | 1306 | 7129657 | ||

Slc7a7
Genomic Map