siRNA Details
Gene Symbol: Slc6a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031343.1 | NP_112633.1 | 2 | 357 | ||
GenBank mRNA | AB021970.1 | - | 2 | 357 | ||
AB021971.1 | - | 2 | 357 | |||
Y13223.1 | - | 2 | 315 |
