siRNA Details
| Gene Symbol: Ptbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031346.1 | NP_112636.1 | 13 | 1592 | ||
| XM_006238206.2 | XP_006238268.1 | 14 | 1680 | |||
| XM_006238207.2 | XP_006238269.1 | 14 | 1672 | |||
| XM_006238208.2 | XP_006238270.1 | 13 | 1637 | |||
| XM_006238209.2 | XP_006238271.1 | 11 | 1284 | |||
| GenBank mRNA | AB023966.1 | - | 13 | 1592 | ||

Ptbp3
Genomic Map