siRNA Details
| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031347.1 | NP_112637.1 | 2 | 174 | ||
| XM_008770219.1 | XP_008768441.1 | 2 | 241 | |||
| GenBank mRNA | AB025784.1 | - | 2 | 174 | ||
| AY237127.1 | - | 2 | 133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppargc1a| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031347.1 | NP_112637.1 | 2 | 174 | ||
| XM_008770219.1 | XP_008768441.1 | 2 | 241 | |||
| GenBank mRNA | AB025784.1 | - | 2 | 174 | ||
| AY237127.1 | - | 2 | 133 | |||