siRNA Details
| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031347.1 | NP_112637.1 | 11 | 2114 | ||
| XM_008770219.1 | XP_008768441.1 | 11 | 2181 | |||
| XM_008770220.1 | XP_008768442.1 | 10 | 3351 | |||
| GenBank mRNA | AB025784.1 | - | 11 | 2114 | ||
| AY237127.1 | - | 11 | 2073 | |||

Ppargc1a
Genomic Map