siRNA Details
| Gene Symbol: Acox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053339.1 | NP_445791.1 | 14 | 1597 | ||
| XM_008770372.1 | XP_008768594.1 | 14 | 4357 | |||
| XM_008770373.1 | XP_008768595.1 | 14 | 4400 | |||
| XM_008770374.1 | XP_008768596.1 | 13 | 1624 | |||
| GenBank mRNA | X95188.1 | - | 14 | 1597 | ||

Acox3
Genomic Map