siRNA Details
| Gene Symbol: Impa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032057.1 | NP_114446.1 | 4 | 343 | ||
| XM_006232151.2 | XP_006232213.1 | 4 | 502 | |||
| GenBank mRNA | U84038.2 | - | 4 | 343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impa1| Gene Symbol: Impa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032057.1 | NP_114446.1 | 4 | 343 | ||
| XM_006232151.2 | XP_006232213.1 | 4 | 502 | |||
| GenBank mRNA | U84038.2 | - | 4 | 343 | ||