siRNA Details
| Gene Symbol: Vdac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031354.1 | NP_112644.1 | 8, 9 | 973 | ||
| XM_006251661.1 | XP_006251723.1 | 9, 10 | 1094 | |||
| GenBank mRNA | AB039663.1 | - | 8, 9 | 973 | ||
| AF268468.1 | - | 7, 8 | 809 | |||
| BC063164.1 | - | 8, 9 | 942 | 6921675 | ||
| FQ214165.1 | - | 5, 6 | 699 | |||
| FQ217146.1 | - | 5, 6 | 698 | |||
| FQ219952.1 | - | 5, 6 | 698 | |||
| FQ228796.1 | - | 5, 6 | 698 | |||

Vdac2
Genomic Map