siRNA Details
Gene Symbol: Vdac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031354.1 | NP_112644.1 | 5 | 394 | ||
XM_006251661.1 | XP_006251723.1 | 6 | 515 | |||
GenBank mRNA | AB039663.1 | - | 5 | 394 | ||
AF268468.1 | - | 4 | 230 | |||
BC063164.1 | - | 5 | 363 | 6921675 | ||
FQ214165.1 | - | 2 | 119 | |||
FQ217146.1 | - | 2 | 119 | |||
FQ219952.1 | - | 2 | 119 | |||
FQ228796.1 | - | 2 | 119 |
