siRNA Details
| Gene Symbol: Aifm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031356.1 | NP_112646.1 | 13 | 1427 | ||
| GenBank mRNA | AB041723.1 | - | 13 | 1443 | ||
| AF262320.1 | - | 13 | 1349 | |||
| AF375656.1 | - | 13 | 1353 | |||
| BC072697.1 | - | 13 | 1385 | 7105761 | ||

Aifm1
Genomic Map