siRNA Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031357.1 | NP_112647.1 | 10, 11 | 1269 | ||
| GenBank mRNA | AB043870.1 | - | 10, 11 | 1269 | ||
| AY724484.1 | - | 10, 11 | 1270 | |||
| BC081775.1 | - | 10, 11 | 1278 | 7107183 | ||

Tpp1
Genomic Map