siRNA Details
| Gene Symbol: Kcnj11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031358.3 | NP_112648.2 | 1 | 350 | ||
| GenBank mRNA | AB043638.1 | - | 1 | 117 | ||
| D61687.1 | - | 1 | 358 | |||
| U44897.1 | - | 1 | 156 | |||
| X97041.1 | - | 1 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj11| Gene Symbol: Kcnj11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031358.3 | NP_112648.2 | 1 | 350 | ||
| GenBank mRNA | AB043638.1 | - | 1 | 117 | ||
| D61687.1 | - | 1 | 358 | |||
| U44897.1 | - | 1 | 156 | |||
| X97041.1 | - | 1 | 114 | |||